siRNA Details
Gene Symbol: Shank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021423.3 | NP_067398.2 | 22 | 5305 | ||
GenBank mRNA | AJ245904.1 | - | 18 | 2878 | ||
AK052445.1 | - | 1 | 1194 | |||
AK173228.1 | - | 2 | 2684 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_021676, AJ133120, AF133301.
Gene Ontology Categories:
