siRNA Details
Gene Symbol: Shank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021423.3 | NP_067398.2 | 22 | 6860 | ||
GenBank mRNA | AJ245904.1 | - | 18 | 4433 | ||
AK052445.1 | - | 1 | 2749 | |||
AK173228.1 | - | 2 | 4239 |

More Information
Other Species Targeted

Gene Ontology Categories:
