siRNA Details
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021433.3 | NP_067408.1 | 2, 3 | 398 | ||
GenBank mRNA | AF237814.1 | - | 2, 3 | 208 | ||
AK009690.1 | - | 2, 3 | 353 | |||
AK018167.1 | - | 2, 3 | 386 | |||
AK019106.1 | - | 2, 3 | 360 | |||
AK031673.1 | - | 2, 3 | 377 | |||
AK044905.1 | - | 2, 3 | 359 | |||
AK049124.1 | - | 2, 3 | 358 | |||
AK151549.1 | - | 2, 3 | 352 | |||
AK159392.1 | - | 2, 3 | 370 | |||
AK179436.1 | - | 1, 2 | 46 | |||
BC029205.1 | - | 2, 3 | 315 | 4039560 | ||
BQ749205.1 | - | 2, 3 | 371 | |||
CJ088919.1 | - | 2, 3 | 402 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031665, BC081769, U56815.
Gene Ontology Categories:
