siRNA Details
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021433.3 | NP_067408.1 | 8 | 1867 | ||
GenBank mRNA | AK019106.1 | - | 8 | 1829 | ||
AK020416.1 | - | 1 | 349 | |||
AK044905.1 | - | 8 | 1828 | |||
AK151549.1 | - | 8 | 1821 | |||
AK191002.1 | - | 1 | 63 | |||
AK198164.1 | - | 1 | 41 | |||
BC029205.1 | - | 8 | 1784 | 4039560 | ||
BU613686.1 | - | 1 | 14 | |||
CF737112.1 | - | 1 | 339 |
