siRNA Details
Gene Symbol: FUT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006581.3 | NP_006572.2 | 3 | 617 | ||
XM_011535383.2 | XP_011533685.1 | 3 | 6354 | |||
XM_011535385.2 | XP_011533687.1 | 4 | 6471 | |||
XM_017010188.1 | XP_016865677.1 | 4 | 3618 | |||
XM_017010190.1 | XP_016865679.1 | 4 | 544 | |||
GenBank mRNA | AB023021.1 | - | 3 | 570 | ||
AJ238701.1 | - | 2 | 380 | |||
AK314966.1 | - | 3 | 612 | |||
AY800264.1 | - | 1 | 276 | |||
BC036101.1 | - | 3 | 594 | 5286309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
