siRNA Details
Gene Symbol: 1700123O20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021437.2 | NP_067412.1 | 2 | 1620 | ||
GenBank mRNA | AB041657.1 | - | 2 | 1575 | ||
AK007261.1 | - | 2 | 1021 | |||
AK167989.1 | - | 2 | 806 | |||
AK170890.1 | - | 2 | 956 | |||
BC011283.1 | - | 2 | 752 | 3581864 |
