siRNA Details
Gene Symbol: 0610007P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021446.2 | NP_067421.1 | 2, 3 | 240 | ||
GenBank mRNA | AF270646.1 | - | 2, 3 | 239 | ||
AK004480.1 | - | 2, 3 | 262 | |||
AK152230.1 | - | 2, 3 | 218 | |||
BC004591.1 | - | 2, 3 | 226 | 3587693 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106749, XM_006240367, XM_006240368, FQ212891.
Gene Ontology Categories:
