siRNA Details
Gene Symbol: Trpm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164325.1 | NP_001157797.1 | 3, 4 | 404 | ||
NM_021450.2 | NP_067425.2 | 3, 4 | 404 | |||
GenBank mRNA | AF149013.1 | - | 3, 4 | 354 | ||
AF376052.1 | - | 3, 4 | 260 | |||
AK014698.1 | - | 3, 4 | 345 | |||
AK052823.1 | - | 3, 4 | 376 | |||
AK162483.1 | - | 3, 4 | 330 | |||
AY032951.1 | - | 3, 4 | 373 | |||
BC145122.1 | - | 3, 4 | 199 | |||
BC150709.1 | - | 3, 4 | 199 | 9087620 |

More Information
Other Species Targeted

Gene Ontology Categories:
