siRNA Details
Gene Symbol: Trpm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164325.1 | NP_001157797.1 | 8, 9 | 1296 | ||
NM_021450.2 | NP_067425.2 | 8, 9 | 1296 | |||
GenBank mRNA | AF149013.1 | - | 8, 9 | 1246 | ||
AF376052.1 | - | 8, 9 | 1152 | |||
AK052823.1 | - | 8, 9 | 1269 | |||
AK162483.1 | - | 8, 9 | 1223 | |||
AY032951.1 | - | 8, 9 | 1265 | |||
BC145122.1 | - | 8, 9 | 1091 | |||
BC150709.1 | - | 8, 9 | 1091 | 9087620 |
