siRNA Details
Gene Symbol: CNPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318842.1 | NP_001305771.1 | 2 | 698 | ||
NM_001318847.1 | NP_001305776.1 | 2 | 698 | |||
NM_001318848.1 | NP_001305777.1 | 2 | 698 | |||
NM_006586.4 | NP_006577.2 | 2 | 698 | |||
NR_134880.1 | 2 | 698 | ||||
NR_134881.1 | 3 | 1891 | ||||
NR_134885.1 | 2 | 795 | ||||
GenBank mRNA | AK090425.1 | - | 1 | 147 | ||
AK124196.1 | - | 3 | 1890 | |||
AY358960.1 | - | 2 | 593 | |||
BC004423.1 | - | 2 | 644 | 2819660 | ||
BC008898.2 | - | 2 | 325 | 2959532 | ||
BC008961.2 | - | 2 | 325 | 2959532 | ||
BG675987.1 | - | 2 | 292 | |||
BQ063472.1 | - | 2 | 304 | |||
BQ680146.1 | - | 2 | 317 | |||
CK429647.1 | - | 2 | 311 | |||
CN302713.1 | - | 2 | 436 | |||
U80744.1 | - | 2 | 644 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
