siRNA Details
Gene Symbol: FAM189B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267608.1 | NP_001254537.1 | 3 | 954 | ||
NM_006589.2 | NP_006580.2 | 4 | 1011 | |||
XM_005244845.1 | XP_005244902.1 | 4 | 1005 | |||
GenBank mRNA | BC006493.2 | - | 4 | 491 | 2966515 | |
BC007444.2 | - | 4 | 491 | 2966515 | ||
BC010471.1 | - | 4 | 915 | 4179576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
