siRNA Details
Gene Symbol: ADORA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000677.3 | NP_000668.1 | 2 | 1645 | ||
NM_001302678.1 | NP_001289607.1 | 2 | 1709 | |||
NM_001302679.1 | NP_001289608.1 | 2 | 1151 | |||
GenBank mRNA | AK313095.1 | - | 2 | 1239 | ||
AY136749.1 | - | 2 | 878 | |||
CD013875.1 | - | 2 | 1000 | |||
JN855714.1 | - | 2 | 877 | |||
L20463.1 | - | 2 | 1169 | |||
L22607.1 | - | 2 | 1143 | |||
X76981.1 | - | 2 | 1211 |
Gene Symbol: LOC105372321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR627135.1 | - | 1 | 1580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
