siRNA Details
Gene Symbol: Thap11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021513.2 | NP_067488.1 | 1 | 1738 | ||
GenBank mRNA | AB041579.1 | - | 1 | 1699 | ||
AK165981.1 | - | 1 | 1699 | |||
AK166109.1 | - | 1 | 1735 | |||
AK170457.1 | - | 1 | 1710 | |||
AK210001.1 | - | 1 | 137 | |||
BC003949.1 | - | 1 | 1291 |
