siRNA Details
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021530.2 | NP_067505.2 | 9, 10 | 1283 | ||
GenBank mRNA | AF224508.1 | - | 9, 10 | 1229 | ||
AK034237.1 | - | 9, 10 | 1285 | |||
AK039404.1 | - | 9, 10 | 1248 | |||
AK047863.1 | - | 2, 3 | 218 | |||
AK171610.1 | - | 8, 9 | 1059 | |||
AK173014.1 | - | 9, 10 | 1117 | |||
AK201792.1 | - | 2, 3 | 142 | |||
BC030388.1 | - | 9, 10 | 1245 | 5401143 |
