siRNA Details
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021530.2 | NP_067505.2 | 10 | 1311 | ||
GenBank mRNA | AF224508.1 | - | 10 | 1257 | ||
AK034237.1 | - | 10 | 1313 | |||
AK039404.1 | - | 10 | 1276 | |||
AK047863.1 | - | 3 | 246 | |||
AK171610.1 | - | 9 | 1087 | |||
AK173014.1 | - | 10 | 1145 | |||
BC030388.1 | - | 10 | 1273 | 5401143 |
