siRNA Details
Gene Symbol: Cope | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021538.1 | NP_067513.1 | 4 | 374 | ||
GenBank mRNA | AB039837.1 | - | 4 | 374 | ||
AK003458.1 | - | 4 | 391 | |||
AK083279.1 | - | 2 | 2069 | |||
AK140753.1 | - | 2 | 2095 | |||
BC009170.1 | - | 4 | 382 | 2812643 | ||
BC057328.1 | - | 2 | 2060 | |||
BC083336.1 | - | 4 | 356 | 5709960 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
