siRNA Details
Gene Symbol: Iigp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146275.1 | NP_001139747.1 | 3 | 788 | ||
NM_021792.4 | NP_068564.4 | 2 | 605 | |||
GenBank mRNA | AF194871.1 | - | 1 | 500 | ||
AJ007971.1 | - | 2 | 587 | |||
AK089702.1 | - | 2 | 616 | |||
AK153060.1 | - | 2 | 607 | |||
BC004649.1 | - | 2 | 582 | 3483102 | ||
CT010300.1 | - | 1 | 483 | |||
DV061261.1 | - | 2 | 638 |
Gene Symbol: Gm4951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033767.3 | NP_001028939.1 | 3 | 702 | ||
GenBank mRNA | AK149556.1 | - | 2 | 594 | ||
AK149583.1 | - | 3 | 700 | |||
BC139145.1 | - | 2 | 530 | 8862167 | ||
BC139147.1 | - | 2 | 530 | 8862169 |
Gene Symbol: Iigp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029756.1 | - | 590 | 4013233 | ||
BC040796.1 | - | 600 | 3495064 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
