siRNA Details
Gene Symbol: Irx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301137.1 | NP_001288066.1 | 5, 6 | 2060 | ||
GenBank mRNA | AJ271055.1 | - | 5, 6 | 1324 | ||
BC036969.1 | - | 2, 3 | 654 | |||
BC130029.1 | - | 5, 6 | 1523 | 40140906 | ||
BU054800.1 | - | 1, 2 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
