siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164207.1 | NP_001157679.1 | 7 | 1355 | ||
NM_001164208.1 | NP_001157680.1 | 7 | 1003 | |||
NM_001164209.2 | NP_001157681.1 | 8 | 1211 | |||
NM_001286651.1 | NP_001273580.1 | 8 | 1208 | |||
NM_001286652.1 | NP_001273581.1 | 7 | 1115 | |||
NM_023056.4 | NP_075543.1 | 7 | 1604 | |||
GenBank mRNA | AB031386.1 | - | 7 | 1164 | ||
AK007408.1 | - | 7 | 1010 | |||
AK156042.1 | - | 7 | 1356 | |||
BC019539.1 | - | 6 | 973 | 4237639 | ||
BC022166.1 | - | 7 | 1619 | 3582123 |
