siRNA Details
Gene Symbol: Tbata | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017407.1 | NP_001017407.1 | 4 | 726 | ||
NM_001017409.1 | NP_001017409.1 | 4 | 793 | |||
NM_001017441.2 | NP_001017441.2 | 5 | 808 | |||
NM_023064.3 | NP_075551.3 | 4 | 828 | |||
GenBank mRNA | AF257502.1 | - | 4 | 828 | ||
AF257503.1 | - | 4 | 726 | |||
AJ249392.1 | - | 3 | 765 | |||
AK038284.1 | - | 3 | 596 | |||
AK088524.1 | - | 3 | 697 | |||
AK132793.1 | - | 5 | 809 | |||
AY243457.1 | - | 4 | 795 | |||
AY243459.1 | - | 3 | 764 | |||
X83884.1 | - | 1 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
