siRNA Details
Gene Symbol: Tbata | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF257502.1 | - | 4 | 938 | ||
AF257503.1 | - | 4 | 836 | |||
AY243457.1 | - | 4 | 905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
