siRNA Details
Gene Symbol: Tmem254c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270498.1 | NP_001257427.1 | 784 | |||
NM_001270499.1 | NP_001257428.1 | 604 | ||||
GenBank mRNA | BY703810.1 | - | 524 | |||
CK128852.1 | - | 699 | ||||
CX201482.1 | - | 603 |
Gene Symbol: Tmem254a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025311.3 | NP_079587.1 | 615 | |||
NM_026679.2 | NP_080955.1 | 784 | ||||
GenBank mRNA | AK002342.1 | - | 4 | 603 | ||
AK002441.1 | - | 4 | 715 | |||
AK003554.1 | - | 4 | 524 | |||
AK008299.1 | - | 4 | 575 | |||
BC031854.1 | - | 4 | 668 | 4194400 |
Gene Symbol: Tmem254b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270495.1 | NP_001257424.1 | 4 | 768 | ||
NM_001270496.1 | NP_001257425.1 | 4 | 526 | |||
GenBank mRNA | CK793021.1 | - | 4 | 524 | ||
DV046977.1 | - | 3 | 386 |
