siRNA Details
Gene Symbol: MTMR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145862.1 | NP_001139334.1 | 3 | 439 | ||
XM_011509099.2 | XP_011507401.1 | 3 | 332 | |||
GenBank mRNA | AK296094.1 | - | 2 | 278 | ||
AK298775.1 | - | 3 | 408 | |||
AK304820.1 | - | 3 | 409 | |||
BC064372.1 | - | 3 | 411 | |||
BC094756.1 | - | 3 | 420 | |||
BC100818.2 | - | 2 | 99 | |||
BC100819.2 | - | 2 | 99 | |||
BC100820.1 | - | 2 | 99 | |||
BC100821.2 | - | 2 | 99 | |||
DA492093.1 | - | 3 | 564 | |||
DB474756.2 | - | 3 | 443 | |||
U78556.1 | - | 3 | 374 | |||
U78557.1 | - | 3 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
