siRNA Details
Gene Symbol: Ndufa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025358.3 | NP_079634.2 | 4 | 408 | ||
GenBank mRNA | AK003137.1 | - | 4 | 390 | ||
AK003214.1 | - | 4 | 381 | |||
AK012366.1 | - | 4 | 388 | |||
BC005760.1 | - | 3 | 289 | |||
BC058378.1 | - | 4 | 376 | 6820787 |
