siRNA Details
Gene Symbol: Tmem14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025387.3 | NP_079663.1 | 6 | 618 | ||
GenBank mRNA | AK003891.1 | - | 6 | 567 | ||
AK008495.1 | - | 5 | 508 | |||
AK008878.1 | - | 6 | 540 | |||
AK010609.1 | - | 6 | 578 | |||
AK012101.1 | - | 6 | 565 | |||
AK019395.1 | - | 6 | 567 | |||
AK050688.1 | - | 6 | 553 | |||
AK089021.1 | - | 5 | 507 | |||
AK211061.1 | - | 2 | 212 | |||
AK217054.1 | - | 2 | 212 | |||
AK219176.1 | - | 2 | 212 | |||
BC025854.1 | - | 6 | 519 | 5250659 | ||
BC055862.1 | - | 6 | 542 | 3492090 | ||
BC069899.1 | - | 5 | 1264 |
