siRNA Details
Gene Symbol: Cd302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290660.1 | NP_001277589.1 | 4 | 729 | ||
NM_025422.4 | NP_079698.2 | 4 | 729 | |||
GenBank mRNA | AK002812.1 | - | 4 | 370 | ||
AK004267.1 | - | 4 | 665 | |||
BC005501.2 | - | 3 | 291 | 3489789 |
