siRNA Details
Gene Symbol: Thoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285780.1 | NP_001272709.1 | 9 | 960 | ||
NM_025435.3 | NP_079711.1 | 8 | 882 | |||
GenBank mRNA | AK005176.1 | - | 8 | 808 | ||
AK010958.1 | - | 8 | 823 | |||
AK147003.1 | - | 9 | 863 | |||
BC027113.1 | - | 7 | 1830 | |||
BC042209.1 | - | 8 | 809 | |||
BC054419.1 | - | 8 | 842 | 4010343 | ||
CF949442.1 | - | 3 | 354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
