siRNA Details
Gene Symbol: Fam134b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034851.2 | NP_001030023.1 | 9 | 1489 | ||
NM_001277315.1 | NP_001264244.1 | 7 | 1578 | |||
NM_001277316.1 | NP_001264245.1 | 7 | 1161 | |||
NM_001277317.1 | NP_001264246.1 | 7 | 1139 | |||
NM_001277318.1 | NP_001264247.1 | 7 | 1111 | |||
NM_025459.3 | NP_079735.2 | 8 | 1279 | |||
GenBank mRNA | AK007506.1 | - | 7 | 1140 | ||
AK015182.1 | - | 8 | 1101 | |||
AK015887.1 | - | 1 | 458 | |||
AK146972.1 | - | 7 | 1162 | |||
AK167327.1 | - | 7 | 1579 | |||
BC019494.1 | - | 8 | 1218 | 4460992 | ||
BC054415.1 | - | 7 | 1098 | 3487542 |
