siRNA Details
Gene Symbol: Senp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012740.1 | - | 3 | 226 | ||
AK013090.1 | - | 3 | 226 | |||
AK083086.1 | - | 3 | 210 | |||
AK085060.1 | - | 3 | 225 | |||
AK122542.1 | - | 3 | 221 | |||
AK164100.1 | - | 3 | 215 | |||
AK164354.1 | - | 3 | 210 | |||
BC058593.1 | - | 3 | 168 | 5364345 | ||
CF583861.1 | - | 4 | 424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
