siRNA Details
Gene Symbol: Senp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083086.1 | - | 16 | 2347 | ||
AK085060.1 | - | 16 | 2361 | |||
AK122542.1 | - | 16 | 2357 | |||
BC058593.1 | - | 16 | 2304 | 5364345 |

More Information
Other Species Targeted

