siRNA Details
Gene Symbol: Zfand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025512.2 | NP_079788.2 | 6 | 454 | ||
GenBank mRNA | AK009240.1 | - | 6 | 468 | ||
AK017696.1 | - | 6 | 468 | |||
AK030472.1 | - | 6 | 462 | |||
AK077873.1 | - | 6 | 464 | |||
BC023352.1 | - | 6 | 446 | |||
BC025216.1 | - | 6 | 446 | |||
BC046585.1 | - | 6 | 454 | 6314252 | ||
BC089494.1 | - | 6 | 466 | 30504018 |
