siRNA Details
Gene Symbol: CSNK2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001896.2 | NP_001887.1 | 12 | 1467 | ||
XM_005255800.3 | XP_005255857.1 | 10 | 1552 | |||
XM_005255801.3 | XP_005255858.1 | 12 | 1463 | |||
XM_017022945.1 | XP_016878434.1 | 8 | 1376 | |||
GenBank mRNA | BC008812.2 | - | 12 | 1341 | 3951914 | |
M55268.1 | - | 13 | 1488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
