siRNA Details
Gene Symbol: Asf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025541.3 | NP_079817.1 | 4 | 2168 | ||
GenBank mRNA | AK031513.1 | - | 4 | 2134 | ||
AK088618.1 | - | 4 | 2111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
