siRNA Details
Gene Symbol: Fam107b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025626.4 | NP_079902.1 | 2 | 309 | ||
GenBank mRNA | AK013932.2 | - | 2 | 274 | ||
AK019815.1 | - | 2 | 221 | |||
AK032875.1 | - | 2 | 310 | |||
AK168759.1 | - | 2 | 275 | |||
BC021353.1 | - | 2 | 275 | 5006672 |
