siRNA Details
Gene Symbol: Fam107b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025626.4 | NP_079902.1 | 4 | 589 | ||
GenBank mRNA | AK013932.2 | - | 4 | 554 | ||
AK019815.1 | - | 4 | 501 | |||
AK032875.1 | - | 4 | 590 | |||
AK168759.1 | - | 4 | 555 | |||
BC021353.1 | - | 4 | 555 | 5006672 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001025034, XM_008771880, XM_008771881, XM_008771883, XM_008771884, and others.
Gene Ontology Categories:
