siRNA Details
Gene Symbol: Teddm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025634.3 | NP_079910.1 | 1 | 1112 | ||
GenBank mRNA | AK009367.1 | - | 1 | 1056 | ||
AK009496.1 | - | 1 | 1069 | |||
AK009566.1 | - | 1 | 1114 | |||
AK009632.1 | - | 1 | 1073 | |||
AK009635.1 | - | 1 | 1074 | |||
AK009751.1 | - | 1 | 1171 | |||
AK009766.1 | - | 1 | 1064 | |||
AK009777.1 | - | 1 | 1056 | |||
AK009984.1 | - | 1 | 1062 | |||
AK009997.1 | - | 1 | 1106 | |||
AK010016.1 | - | 1 | 1073 | |||
BC115655.1 | - | 1 | 1063 | 40103183 | ||
BC115656.1 | - | 1 | 1062 | 40103184 |
