siRNA Details
Gene Symbol: Crls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024385.1 | NP_001019556.1 | 2, 3 | 599 | ||
NM_025646.3 | NP_079922.2 | 2, 3 | 286 | |||
GenBank mRNA | AK002408.1 | - | 2, 3 | 601 | ||
AK004215.1 | - | 2, 3 | 315 | |||
AK016165.1 | - | 2, 3 | 287 | |||
AK018180.1 | - | 2, 3 | 600 | |||
BC048702.1 | - | 2, 3 | 580 | 6506241 | ||
BC086619.1 | - | 2, 3 | 558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
