siRNA Details
Gene Symbol: Ms4a4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025658.4 | NP_079934.2 | 5, 6 | 769 | ||
GenBank mRNA | AF237915.1 | - | 4, 5 | 443 | ||
AK003110.1 | - | 5, 6 | 811 | |||
AK008968.1 | - | 5, 6 | 818 | |||
BC069842.1 | - | 5, 6 | 769 | 30446709 |
