siRNA Details
Gene Symbol: Sltm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025690.3 | NP_079966.2 | 21 | 3543 | ||
GenBank mRNA | AF462146.1 | - | 21 | 3543 | ||
AK134756.1 | - | 12 | 2222 | |||
BC019992.1 | - | 10 | 1913 | |||
CN527065.1 | - | 3 | 717 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
