siRNA Details
Gene Symbol: Rbm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025717.3 | NP_079993.2 | 3 | 806 | ||
GenBank mRNA | AK006447.1 | - | 3 | 789 | ||
AK142116.1 | - | 3 | 762 | |||
AK160796.1 | - | 3 | 1201 | |||
AK181241.1 | - | 1 | 213 | |||
AK182975.1 | - | 1 | 213 | |||
BC019488.1 | - | 3 | 711 | 4457936 |
