siRNA Details
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025730.3 | NP_080006.3 | 51 | 8109 | ||
GenBank mRNA | AK014886.1 | - | 12 | 2168 | ||
AK050702.1 | - | 2 | 696 | |||
AK156100.1 | - | 4 | 3782 | |||
BC034074.1 | - | 17 | 2992 | |||
BC035949.1 | - | 7 | 1453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
