siRNA Details
Gene Symbol: Ankrd61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025732.3 | NP_080008.1 | 3 | 413 | ||
GenBank mRNA | AK014929.1 | - | 3 | 414 | ||
AK017053.1 | - | 3 | 354 | |||
AK146023.1 | - | 692 | ||||
BC052346.1 | - | 3 | 635 | 6703467 |
