siRNA Details
Gene Symbol: SRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006662.2 | NP_006653.2 | 8 | 1284 | ||
GenBank mRNA | AB621816.1 | - | 8 | 1351 | ||
AF143946.1 | - | 4 | 518 | |||
AK128030.1 | - | 8 | 1284 | |||
BC025284.1 | - | 5 | 761 | |||
BC063578.1 | - | 6 | 1078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
