siRNA Details
Gene Symbol: 4933427E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006089.1 | - | 1 | 602 | ||
AK006196.1 | - | 1 | 577 | |||
AK006985.1 | - | 1 | 599 | |||
AK016947.1 | - | 1 | 789 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933427E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006089.1 | - | 1 | 602 | ||
AK006196.1 | - | 1 | 577 | |||
AK006985.1 | - | 1 | 599 | |||
AK016947.1 | - | 1 | 789 |