siRNA Details
Gene Symbol: 4933436I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025763.1 | NP_080039.1 | 2 | 340 | ||
GenBank mRNA | AK016292.1 | - | 2 | 324 | ||
AK017084.1 | - | 2 | 340 | |||
BC125317.1 | - | 2 | 326 | 40129832 | ||
BC138016.1 | - | 2 | 326 | 8861035 |
