siRNA Details
Gene Symbol: LTB4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC004545.2 | - | 2 | 1536 | 3946189 | |
D89078.1 | - | 2 | 2921 | |||
D89079.1 | - | 2 | 1525 | |||
U41070.1 | - | 2 | 1508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
