siRNA Details
Gene Symbol: Ccdc109b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293644.1 | NP_001280573.1 | 1, 2 | 265 | ||
NM_025779.3 | NP_080055.2 | 1, 2 | 265 | |||
GenBank mRNA | AK018492.1 | - | 1, 2 | 142 | ||
AK150724.1 | - | 1, 2 | 163 | |||
AK150995.1 | - | 1, 2 | 139 | |||
AK151201.1 | - | 1, 2 | 163 | |||
AK151915.1 | - | 1, 2 | 176 | |||
AK152326.1 | - | 1, 2 | 136 | |||
AK154338.1 | - | 1, 2 | 158 | |||
BC049571.1 | - | 1, 2 | 265 | 6588325 |
