siRNA Details
Gene Symbol: Ccdc109b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293644.1 | NP_001280573.1 | 5, 6 | 814 | ||
NM_025779.3 | NP_080055.2 | 6, 7 | 975 | |||
GenBank mRNA | AK013032.1 | - | 2, 3 | 283 | ||
AK018492.1 | - | 6, 7 | 852 | |||
AK150724.1 | - | 6, 7 | 873 | |||
AK150995.1 | - | 6, 7 | 849 | |||
AK151201.1 | - | 6, 7 | 873 | |||
AK151915.1 | - | 6, 7 | 886 | |||
AK152326.1 | - | 5, 6 | 685 | |||
AK154338.1 | - | 6, 7 | 868 | |||
BC049571.1 | - | 6, 7 | 975 | 6588325 | ||
BX515874.1 | - | 1, 2 | 26 |
