siRNA Details
Gene Symbol: Ppp1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025800.3 | NP_080076.1 | 5 | 729 | ||
GenBank mRNA | AK002671.1 | - | 5 | 676 | ||
AK004320.1 | - | 4 | 299 | |||
AK028603.1 | - | 5 | 681 | |||
AK032462.1 | - | 5 | 680 | |||
AK053657.1 | - | 5 | 630 | |||
AK150281.1 | - | 5 | 732 | |||
AK207871.1 | - | 4 | 246 | |||
BC069885.1 | - | 5 | 702 | 3671082 | ||
BC069886.1 | - | 5 | 674 | 6310133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
