siRNA Details
Gene Symbol: Tcf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037877.3 | NP_001032966.1 | 18 | 2106 | ||
NM_001286362.1 | NP_001273291.1 | 17 | 2028 | |||
NM_025804.3 | NP_080080.2 | 18 | 2103 | |||
GenBank mRNA | AK003180.1 | - | 18 | 1993 | ||
AK007744.1 | - | 1 | 103 | |||
AK013754.1 | - | 18 | 1792 | |||
AK045397.1 | - | 17 | 1957 | |||
AK050274.1 | - | 18 | 2029 | |||
AK075728.1 | - | 18 | 1953 | |||
AK076151.1 | - | 12 | 1193 | |||
AK088602.1 | - | 5 | 642 | |||
AK089964.1 | - | 18 | 1995 | |||
AK122430.1 | - | 17 | 4410 | |||
AK148082.1 | - | 19 | 1968 | |||
AK161627.1 | - | 18 | 1982 | |||
BC016130.1 | - | 18 | 1968 | |||
BC046768.1 | - | 18 | 2061 | 5707312 | ||
U94988.1 | - | 18 | 1964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
